Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PGAP2 SIRNA Assays ›
Gene Symbol:
PGAP2
Gene Name:
post-GPI attachment to proteins 2
Gene Aliases:
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location:
Chr.11: 3797724 - 3826371 on Build GRCh38
UniGene ID:
Hs.133968
Species:
Human
siRNA ID
HSS178420
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PGAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145438.2
NP_001138910.1
6
803
NM_001256235.1
NP_001243164.1
7
756
NM_001256236.1
NP_001243165.1
7
998
NM_001256237.1
NP_001243166.1
6
894
NM_001256238.1
NP_001243167.1
6
802
NM_001256239.1
NP_001243168.1
5
733
NM_001256240.1
NP_001243169.1
5
745
NM_001283038.1
NP_001269967.1
6
815
NM_001283039.1
NP_001269968.1
6
768
NM_001283040.1
NP_001269969.1
4
387
NM_014489.3
NP_055304.1
6
928
NR_027016.2
4
570
NR_027017.2
7
1102
NR_027018.2
7
767
NR_045923.1
5
640
NR_045925.1
5
550
NR_045926.1
5
562
NR_045927.1
5
719
NR_045929.1
5
686
NR_104270.1
5
628
NR_104271.1
7
888
NR_104272.1
4
558
XM_006718181.3
XP_006718244.1
5
1406
XM_006718183.3
XP_006718246.1
7
823
XM_006718185.2
XP_006718248.1
6
715
XM_006718186.2
XP_006718249.1
7
875
XM_006718190.3
XP_006718253.1
6
1031
XM_006718191.3
XP_006718254.1
6
1019
XM_006718192.1
XP_006718255.1
8
949
XM_006718193.2
XP_006718256.1
7
756
XM_011519990.2
XP_011518292.2
7
1282
XM_011519991.2
XP_011518293.2
7
1273
XM_011519992.1
XP_011518294.1
8
1054
XM_011519993.2
XP_011518295.2
7
1380
XM_011519994.2
XP_011518296.2
7
1374
XM_011519995.2
XP_011518297.1
7
1205
XM_011519996.1
XP_011518298.1
8
1006
XM_011519997.1
XP_011518299.1
8
1347
XM_011519998.2
XP_011518300.1
8
1025
XM_011519999.1
XP_011518301.1
8
984
XM_011520000.1
XP_011518302.1
7
898
XM_011520001.1
XP_011518303.1
6
690
XM_011520002.1
XP_011518304.1
6
678
XM_011520003.2
XP_011518305.1
7
1214
XM_011520004.2
XP_011518306.1
7
889
XM_011520005.2
XP_011518307.1
6
786
XM_011520006.2
XP_011518308.1
7
1170
XM_011520007.2
XP_011518309.1
8
939
XM_017017556.1
XP_016873045.1
6
769
XM_017017557.1
XP_016873046.1
6
838
XM_017017558.1
XP_016873047.1
6
1012
XM_017017559.1
XP_016873048.1
7
875
XM_017017560.1
XP_016873049.1
7
863
GenBank mRNA
AF159615.1
-
6
930
AK292181.1
-
7
892
AK295202.1
-
7
767
AK301991.1
-
7
1072
AK307359.1
-
5
557
AL049261.1
-
2
1957
AL096753.1
-
7
756
AY373030.1
-
4
619
BC009930.2
-
6
690
3506614
BG468212.1
-
5
439
BG707484.1
-
5
613
BI768071.1
-
5
702
BI858175.1
-
1
87
BX340489.2
-
5
670
BX356273.2
-
6
722
BX379511.2
-
6
770
BX448155.2
-
6
674
CB988453.1
-
4
585
CD675009.1
-
3
281
CD675113.1
-
5
521
CX872317.1
-
4
498
HY171112.1
-
4
308
KU178608.1
-
4
607
KU178609.1
-
5
777
KU178610.1
-
5
704
KU178611.1
-
4
619
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
protein transport
signal transduction in response to DNA damage
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-z4fcc:80/10.239.160.202:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0