Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH11X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168360.1 | NP_001161832.1 | 3 | 3120 | ||
NM_001168361.1 | NP_001161833.1 | 3 | 3120 | |||
NM_001168363.1 | NP_001161835.1 | 3 | 3120 | |||
NM_032968.4 | NP_116750.1 | 3 | 3120 | |||
NM_032969.4 | NP_116751.1 | 3 | 3120 | |||
XM_011530910.1 | XP_011529212.1 | 6 | 3595 | |||
XM_011530911.2 | XP_011529213.1 | 8 | 3883 | |||
XM_011530912.2 | XP_011529214.1 | 7 | 3829 | |||
XM_011530913.2 | XP_011529215.1 | 7 | 3829 | |||
XM_011530914.2 | XP_011529216.1 | 4 | 3495 | |||
XM_017029416.1 | XP_016884905.1 | 3 | 3104 | |||
XM_017029417.1 | XP_016884906.1 | 7 | 3829 | |||
XM_017029418.1 | XP_016884907.1 | 7 | 3829 | |||
XM_017029421.1 | XP_016884910.1 | 3 | 3104 | |||
GenBank mRNA | AB037747.1 | - | 2 | 308 | ||
AF206516.1 | - | 3 | 3076 | |||
AF332218.1 | - | 3 | 3921 | |||
AF332219.1 | - | 3 | 3921 | |||
AJ564938.1 | - | 1 | 43 | |||
AJ564939.1 | - | 1 | 43 | |||
AY861432.1 | - | 3 | 3076 | |||
AY861434.1 | - | 3 | 3076 | |||
AY861435.1 | - | 3 | 3076 |