Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GNAS SIRNA Assays ›
Gene Symbol:
GNAS
Gene Name:
GNAS complex locus
Gene Aliases:
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
Chromosome Location:
Chr.20: 58839681 - 58911196 on Build GRCh38
UniGene ID:
Hs.125898
Species:
Human
siRNA ID
HSS178474
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GNAS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000516.5
NP_000507.1
2
607
NM_001077488.3
NP_001070956.1
2
607
NM_001077489.3
NP_001070957.1
2
607
NM_001077490.2
NP_001070958.1
2
2397
NM_001309840.1
NP_001296769.1
2
273
NM_001309842.1
NP_001296771.1
2
607
NM_001309861.1
NP_001296790.1
2
318
NM_001309883.1
NP_001296812.1
4
2555
NM_016592.3
NP_057676.1
2
1213
NM_080425.3
NP_536350.2
2
2397
NM_080426.3
NP_536351.1
2
607
NR_132272.1
2
90
NR_132273.1
2
450
XM_017027812.1
XP_016883301.1
2
2664
XM_017027813.1
XP_016883302.1
2
2664
XM_017027814.1
XP_016883303.1
2
2664
XM_017027815.1
XP_016883304.1
2
135
XM_017027816.1
XP_016883305.1
2
813
XM_017027817.1
XP_016883306.1
2
222
XM_017027818.1
XP_016883307.1
2
145
XM_017027819.1
XP_016883308.1
2
186
XM_017027820.1
XP_016883309.1
1
44
XM_017027821.1
XP_016883310.1
2
1250
XM_017027822.1
XP_016883311.1
2
1250
GenBank mRNA
AF105253.1
-
2
1191
AF493897.1
-
2
183
AF493898.1
-
2
183
AJ224867.1
-
4
578
AK026564.1
-
3
377
AK093534.1
-
1
459
AK225818.1
-
3
351
AK315860.1
-
2
105
AK315874.1
-
2
286
AL537118.3
-
2
303
AV751821.1
-
2
450
BC002722.2
-
3
301
BC008855.2
-
3
325
BC022875.1
-
2
74
BC066923.1
-
2
186
BC089157.1
-
2
499
BC104928.1
-
3
271
BC108315.1
-
2
448
BM894355.1
-
2
148
BT009905.1
-
2
183
BU857975.1
-
2
497
CN295544.1
-
2
271
CN403086.1
-
2
231
DB483472.1
-
2
186
HY141824.1
-
2
90
KU178095.1
-
2
183
KU178096.1
-
2
183
M14631.1
-
2
150
X04408.1
-
2
251
X04409.1
-
2
195
X07036.1
-
2
195
X56009.1
-
2
259
Back To Top
More Information
Panther Classification:
Molecular Function -
heterotrimeric G-protein
Gene Ontology Categories:
Function(s)
Process(es)
tissue homeostasis
endochondral ossification
energy reserve metabolic process
DNA methylation
adenylate cyclase-activating G-protein coupled receptor signaling pathway
adenylate cyclase-activating dopamine receptor signaling pathway
sensory perception of chemical stimulus
embryonic hindlimb morphogenesis
multicellular organism growth
post-embryonic body morphogenesis
response to drug
positive regulation of osteoblast differentiation
positive regulation of osteoclast differentiation
developmental growth
embryonic cranial skeleton morphogenesis
cognition
cartilage development
bone development
hair follicle placode formation
platelet aggregation
genetic imprinting
regulation of parathyroid hormone secretion
GTPase activity
signal transducer activity
insulin-like growth factor receptor binding
protein binding
GTP binding
G-protein beta/gamma-subunit complex binding
beta-2 adrenergic receptor binding
D1 dopamine receptor binding
mu-type opioid receptor binding
ionotropic glutamate receptor binding
metal ion binding
corticotropin-releasing hormone receptor 1 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0