Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000175.3 | NP_000166.2 | 6 | 654 | ||
NM_001184722.1 | NP_001171651.1 | 6 | 717 | |||
NM_001289789.1 | NP_001276718.1 | 7 | 801 | |||
NM_001289790.1 | NP_001276719.1 | 5 | 570 | |||
XM_005258764.2 | XP_005258821.1 | 7 | 620 | |||
XM_006723148.2 | XP_006723211.1 | 7 | 571 | |||
XM_011526754.2 | XP_011525056.1 | 8 | 1036 | |||
GenBank mRNA | AK129884.1 | - | 7 | 585 | ||
AK130102.1 | - | 5 | 349 | |||
AK293446.1 | - | 5 | 446 | |||
AK294396.1 | - | 6 | 665 | |||
AK301103.1 | - | 6 | 488 | |||
BC004982.1 | - | 6 | 522 | 2906270 | ||
K03515.1 | - | 6 | 529 |