Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAJIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037225.2 | NP_001032302.1 | 6 | 524 | ||
NM_001318808.1 | NP_001305737.1 | 6 | 1238 | |||
XM_005273918.2 | XP_005273975.1 | 6 | 518 | |||
XM_006718502.2 | XP_006718565.1 | 6 | 518 | |||
XM_006718503.3 | XP_006718566.1 | 6 | 404 | |||
XM_006718504.2 | XP_006718567.1 | 6 | 518 | |||
XM_011544940.2 | XP_011543242.1 | 6 | 399 | |||
XM_011544941.2 | XP_011543243.1 | 1 | 249 | |||
GenBank mRNA | AK093130.1 | - | 6 | 1232 | ||
BC106951.2 | - | 5 | 319 | 40035116 | ||
BC106952.1 | - | 5 | 319 | 40035117 | ||
DB445650.2 | - | 6 | 525 | |||
LC068587.1 | - | 4 | 245 |