Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173582.4 | NP_775853.2 | 12, 13 | 1940 | ||
XM_011544953.2 | XP_011543255.1 | 13, 14 | 1995 | |||
GenBank mRNA | AB019210.1 | - | 12, 13 | 1891 | ||
AK056591.1 | - | 12, 13 | 1898 | |||
BC059360.1 | - | 12, 13 | 1904 | 30332467 |