Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135919.1 | NP_001129391.1 | 5 | 1761 | ||
NM_181785.3 | NP_861450.1 | 5 | 1761 | |||
XM_005266361.2 | XP_005266418.1 | 5 | 1439 | |||
GenBank mRNA | AF315594.1 | - | 5 | 1345 | ||
AK026720.1 | - | 1 | 540 | |||
AK093932.1 | - | 4 | 2189 | |||
AK098426.1 | - | 3 | 791 | |||
AK124604.1 | - | 3 | 2454 | |||
BC036662.1 | - | 5 | 1723 | 4827143 | ||
BC060850.1 | - | 5 | 1582 | 4820254 | ||
BC068556.1 | - | 5 | 1589 | 4820792 | ||
BX537565.1 | - | 5 | 1572 | |||
CB851220.1 | - | 3 | 288 |