Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213601.1 | NP_998766.1 | 3, 4 | 325 | ||
XM_017021225.1 | XP_016876714.1 | 3, 4 | 2786 | |||
GenBank mRNA | AK095650.1 | - | 3, 4 | 369 | ||
AY598329.1 | - | 3, 4 | 325 | |||
BC101510.1 | - | 3, 4 | 381 | 8069016 |