Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB561D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134400.1 | NP_001127872.1 | 3 | 791 | ||
NM_001134402.1 | NP_001127874.1 | 2 | 597 | |||
NM_001134403.1 | NP_001127875.1 | 4 | 829 | |||
NM_001134404.1 | NP_001127876.1 | 4 | 906 | |||
NM_182580.2 | NP_872386.1 | 3 | 725 | |||
XM_005270775.2 | XP_005270832.1 | 3 | 802 | |||
XM_005270776.3 | XP_005270833.1 | 3 | 641 | |||
XM_005270777.2 | XP_005270834.1 | 2 | 681 | |||
XM_011541286.2 | XP_011539588.1 | 4 | 790 | |||
XM_011541287.2 | XP_011539589.2 | 3 | 1918 | |||
XM_017001078.1 | XP_016856567.1 | 4 | 713 | |||
XM_017001079.1 | XP_016856568.1 | 3 | 535 | |||
GenBank mRNA | AK096354.1 | - | 3 | 704 | ||
AK126707.1 | - | 3 | 802 | |||
AK128798.1 | - | 4 | 870 | |||
AK294992.1 | - | 3 | 756 | |||
AK299086.1 | - | 3 | 591 | |||
AK303546.1 | - | 2 | 666 | |||
BC093683.1 | - | 3 | 699 | 7939528 | ||
BC111999.1 | - | 3 | 699 | 8327467 | ||
BC143308.1 | - | 2 | 661 | |||
BX647509.1 | - | 4 | 809 |