Gene Symbol: EPHA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099439.1 | NP_001092909.1 | 2 | 240 | ||
NM_173641.2 | NP_775912.2 | 2 | 240 | |||
GenBank mRNA | AJ781169.1 | - | 2 | 240 | ||
AK090974.1 | - | 2 | 221 | |||
BC067734.1 | - | 2 | 240 | 4375803 |