Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085454.1 | NP_001078923.1 | 6, 7 | 940 | ||
NM_014030.3 | NP_054749.2 | 6, 7 | 940 | |||
XM_011524684.1 | XP_011522986.1 | 6, 7 | 781 | |||
XM_011524685.1 | XP_011522987.1 | 6, 7 | 781 | |||
GenBank mRNA | AB209537.1 | - | 6, 7 | 739 | ||
AF124490.1 | - | 6, 7 | 740 | |||
AK024826.1 | - | 1, 2 | 95 | |||
AK294785.1 | - | 7, 8 | 818 | |||
AK299932.1 | - | 6, 7 | 784 | |||
BC067358.1 | - | 6, 7 | 940 | |||
BQ424386.1 | - | 5, 6 | 579 |