Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013242.2 | NP_037374.1 | 4 | 708 | ||
NR_136632.1 | 4 | 741 | ||||
GenBank mRNA | AF087845.1 | - | 4 | 688 | ||
AF093680.1 | - | 4 | 687 | |||
AK027099.1 | - | 4 | 629 | |||
AK297819.1 | - | 4 | 663 | |||
BC005152.2 | - | 4 | 677 | 3528301 | ||
BU730963.1 | - | 3 | 603 | |||
CN364433.1 | - | 4 | 668 |