Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GSN SIRNA Assays ›
Gene Symbol:
GSN
Gene Name:
gelsolin
Gene Aliases:
ADF, AGEL
Chromosome Location:
Chr.9: 121201082 - 121332844 on Build GRCh38
UniGene ID:
Hs.522373
Species:
Human
siRNA ID
HSS179025
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GSN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000177.4
NP_000168.1
10
1470
NM_001127662.1
NP_001121134.1
10
1383
NM_001127663.1
NP_001121135.2
12
1588
NM_001127664.1
NP_001121136.1
12
1555
NM_001127665.1
NP_001121137.1
12
1550
NM_001127666.1
NP_001121138.1
12
1547
NM_001127667.1
NP_001121139.1
12
1538
NM_001258029.1
NP_001244958.1
10
1370
NM_001258030.1
NP_001244959.1
10
1447
NM_198252.2
NP_937895.1
11
1476
XM_005251943.1
XP_005252000.1
11
1574
XM_005251944.1
XP_005252001.1
11
1565
XM_005251945.3
XP_005252002.1
11
1478
XM_006717079.1
XP_006717142.1
11
1412
XM_011518584.1
XP_011516886.1
12
1565
XM_011518585.1
XP_011516887.1
10
1385
XM_011518586.1
XP_011516888.1
13
1588
XM_011518587.2
XP_011516889.1
13
1630
XM_011518588.2
XP_011516890.1
13
1592
XM_011518589.2
XP_011516891.1
13
1588
XM_011518590.2
XP_011516892.1
13
1540
XM_011518591.2
XP_011516893.1
18
2632
XM_011518592.1
XP_011516894.1
12
1454
XM_011518593.1
XP_011516895.1
13
1600
XM_011518594.1
XP_011516896.1
7
797
XM_017014643.1
XP_016870132.1
19
2783
XM_017014644.1
XP_016870133.1
19
2717
XM_017014645.1
XP_016870134.1
11
1529
XM_017014646.1
XP_016870135.1
14
1700
XM_017014647.1
XP_016870136.1
20
2916
XM_017014648.1
XP_016870137.1
13
1527
GenBank mRNA
AK092797.1
-
6
1611
AK096280.1
-
11
1449
AK125819.1
-
19
2279
AK295201.1
-
11
1314
AK295572.1
-
12
1509
AK297870.1
-
10
1328
AK299453.1
-
10
1435
AK300631.1
-
12
1522
AK304629.1
-
10
1391
AK304675.1
-
9
1424
AK307859.1
-
10
1334
AK315494.1
-
10
1441
BC026033.1
-
10
1414
4661084
BX647999.1
-
10
1372
X04412.1
-
10
1423
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis, engulfment
aging
positive regulation of gene expression
oligodendrocyte development
striated muscle atrophy
extracellular matrix disassembly
actin filament polymerization
protein destabilization
tissue regeneration
sequestering of actin monomers
cellular protein metabolic process
actin nucleation
response to ethanol
negative regulation of viral entry into host cell
phosphatidylinositol-mediated signaling
actin filament severing
barbed-end actin filament capping
positive regulation of actin nucleation
response to folic acid
actin filament capping
cilium morphogenesis
cellular response to cadmium ion
regulation of podosome assembly
actin filament reorganization
renal protein absorption
hepatocyte apoptotic process
positive regulation of keratinocyte apoptotic process
regulation of wound healing, spreading of epidermal cells
regulation of establishment of T cell polarity
regulation of plasma membrane raft polarization
regulation of receptor clustering
positive regulation of protein processing in phagocytic vesicle
amyloid fibril formation
positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
actin binding
calcium ion binding
protein binding
protein domain specific binding
myosin II binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0