Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323627.1 | NP_001310556.1 | 4 | 686 | ||
NM_013304.2 | NP_037436.1 | 4 | 628 | |||
XM_005255915.1 | XP_005255972.1 | 3 | 362 | |||
XM_006721185.1 | XP_006721248.1 | 3 | 362 | |||
XM_011523058.1 | XP_011521360.1 | 3 | 362 | |||
XM_011523059.1 | XP_011521361.1 | 3 | 362 | |||
XM_011523060.1 | XP_011521362.1 | 2 | 161 | |||
GenBank mRNA | BC021908.1 | - | 4 | 654 | 4053374 | |
U90653.2 | - | 4 | 628 |