Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ERO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014584.2 | NP_055399.1 | 16 | 1840 | ||
XM_011536684.2 | XP_011534986.1 | 15 | 1647 | |||
GenBank mRNA | AF081886.1 | - | 16 | 1616 | ||
AF123887.1 | - | 14 | 1142 | |||
AK292839.1 | - | 16 | 1438 | |||
AY358463.1 | - | 16 | 1562 | |||
BC008674.1 | - | 16 | 1612 | 3866930 | ||
BC012941.1 | - | 16 | 1623 | 3868538 |