Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFEMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016938.4 | NP_058634.4 | 7 | 966 | ||
NR_037718.1 | 7 | 966 | ||||
GenBank mRNA | AB030655.1 | - | 7 | 803 | ||
AB209121.1 | - | 4 | 3042 | |||
AF093119.1 | - | 6 | 695 | |||
AF109121.1 | - | 7 | 794 | |||
AF124486.1 | - | 6 | 707 | |||
AJ132819.2 | - | 6 | 707 | |||
AK000980.1 | - | 7 | 803 | |||
AK075453.1 | - | 7 | 772 | |||
AK098186.1 | - | 3 | 1706 | |||
AK292079.1 | - | 7 | 811 | |||
AK315181.1 | - | 7 | 772 | |||
AY358899.1 | - | 7 | 768 | |||
BC010456.1 | - | 7 | 822 | 4182121 | ||
BC018871.2 | - | 7 | 843 | |||
BC109225.1 | - | 3 | 694 | |||
BQ478413.1 | - | 4 | 386 | |||
CR541934.1 | - | 6 | 707 |