Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000700.2 | NP_000691.1 | 4 | 406 | ||
XM_011518609.1 | XP_011516911.1 | 4 | 333 | |||
XM_017014657.1 | XP_016870146.1 | 4 | 352 | |||
GenBank mRNA | AB451274.1 | - | 3 | 198 | ||
AB451401.1 | - | 3 | 198 | |||
AK074480.1 | - | 3 | 1080 | |||
AK296808.1 | - | 4 | 272 | |||
AK311684.1 | - | 3 | 88 | |||
BC001275.1 | - | 4 | 250 | 3459615 | ||
BC035993.1 | - | 4 | 272 | 4662939 | ||
BT019896.1 | - | 3 | 198 | |||
BT019916.1 | - | 3 | 198 | |||
BT019917.1 | - | 3 | 198 | |||
CR407684.1 | - | 3 | 198 | |||
CX870105.1 | - | 4 | 406 | |||
X05908.1 | - | 4 | 272 |