Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004965.6 | NP_004956.5 | 4, 5 | 328 | ||
XM_017028338.1 | XP_016883827.1 | 4, 5 | 321 | |||
GenBank mRNA | AK315410.1 | - | 4, 5 | 271 | ||
AL582106.2 | - | 2, 3 | 943 | |||
BC000075.2 | - | 4, 5 | 248 | 3510312 | ||
BC023984.1 | - | 4, 5 | 240 | 4104558 | ||
BC070153.1 | - | 4, 5 | 266 | 6450636 | ||
BC070154.1 | - | 4, 5 | 246 | 6495327 | ||
BC071643.1 | - | 4, 5 | 236 | 5549431 | ||
BC072672.1 | - | 4, 5 | 237 | 6166317 | ||
BC106080.1 | - | 4, 5 | 261 | 3855939 | ||
BC107741.1 | - | 4, 5 | 334 | 6495117 | ||
BE779939.1 | - | 387 | ||||
BT007337.1 | - | 1 | 124 | |||
J02621.1 | - | 4, 5 | 274 |
Gene Symbol: CPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC046360.1 | - | 4 | 1213 |