Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127204.1 | NP_001120676.1 | 7 | 1239 | ||
NM_001127205.1 | NP_001120677.1 | 6 | 1143 | |||
NM_001127206.2 | NP_001120678.1 | 6 | 1118 | |||
NM_001286267.1 | NP_001273196.1 | 7 | 1242 | |||
NM_001286268.1 | NP_001273197.1 | 6 | 1299 | |||
NM_001286269.1 | NP_001273198.1 | 7 | 1220 | |||
NM_001286270.1 | NP_001273199.1 | 8 | 1519 | |||
NM_001286271.1 | NP_001273200.1 | 5 | 994 | |||
NM_002134.3 | NP_002125.3 | 6 | 1121 | |||
XM_011522473.1 | XP_011520775.1 | 6 | 1109 | |||
XM_017023196.1 | XP_016878685.1 | 7 | 1163 | |||
XM_017023197.1 | XP_016878686.1 | 6 | 994 | |||
GenBank mRNA | AK093388.1 | - | 8 | 1480 | ||
AK222629.1 | - | 6 | 1013 | |||
AK313660.1 | - | 6 | 1071 | |||
BC002396.2 | - | 6 | 1037 | 2821444 | ||
BT019788.1 | - | 5 | 932 | |||
CR456760.1 | - | 5 | 932 | |||
CR541839.1 | - | 5 | 932 | |||
D21243.1 | - | 5 | 932 | |||
S34389.1 | - | 6 | 1007 |