Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181532.3 | NP_853510.1 | 2 | 412 | ||
GenBank mRNA | AB093575.1 | - | 1 | 583 | ||
BC101642.1 | - | 1 | 352 | 8069148 | ||
BC101644.1 | - | 1 | 352 | 8069150 | ||
HY043532.1 | - | 2 | 413 |