Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001541.3 | NP_001532.1 | 2 | 458 | ||
GenBank mRNA | AB096250.1 | - | 2 | 952 | ||
AI131288.1 | - | 1 | 397 | |||
BC109393.1 | - | 2 | 393 | 40035056 | ||
CR457408.1 | - | 2 | 364 | |||
D89617.1 | - | 2 | 433 | |||
S67070.1 | - | 1 | 103 |
Gene Symbol: HSPB2-C11orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK293041.1 | - | 2 | 458 |