Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000873.3 | NP_000864.2 | 3 | 577 | ||
NM_001099786.1 | NP_001093256.1 | 5 | 700 | |||
NM_001099787.1 | NP_001093257.1 | 4 | 618 | |||
NM_001099788.1 | NP_001093258.1 | 5 | 583 | |||
NM_001099789.1 | NP_001093259.1 | 4 | 501 | |||
GenBank mRNA | AB209878.1 | - | 4 | 609 | ||
AF340052.1 | - | 3 | 382 | |||
AF340053.1 | - | 3 | 391 | |||
AK223219.1 | - | 3 | 421 | |||
AK293110.1 | - | 3 | 423 | |||
BC003097.1 | - | 3 | 399 | 3502827 | ||
BM695682.1 | - | 3 | 577 | |||
BX363874.2 | - | 5 | 645 | |||
CR541812.1 | - | 3 | 345 | |||
CR541834.1 | - | 3 | 345 | |||
X15606.1 | - | 3 | 407 |
Gene Symbol: PRR29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164257.1 | NP_001157729.1 | 6 | 2767 | ||
NM_001191029.1 | NP_001177958.1 | 5 | 2694 | |||
NM_001191030.1 | NP_001177959.1 | 6 | 2746 | |||
NM_001191031.1 | NP_001177960.1 | 5 | 2540 | |||
XM_011525469.1 | XP_011523771.1 | 5 | 2685 | |||
XM_017025315.1 | XP_016880804.1 | 4 | 2421 | |||
GenBank mRNA | AK021786.1 | - | 6 | 2765 | ||
AL161972.1 | - | 2 | 2264 | |||
BC045676.1 | - | 4 | 2719 |