Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178562.4 | NP_848657.1 | 6, 7 | 855 | ||
XM_006715960.3 | XP_006716023.1 | 6, 7 | 683 | |||
GenBank mRNA | AF276891.1 | - | 6, 7 | 590 | ||
AK000208.1 | - | 1, 2 | 469 | |||
AY236849.1 | - | 6, 7 | 679 | |||
BC044244.1 | - | 6, 7 | 819 | 5759132 |