Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000596.2 | NP_000587.1 | 3, 4 | 945 | ||
GenBank mRNA | AK314925.1 | - | 3, 4 | 817 | ||
AV695706.2 | - | 3, 4 | 268 | |||
BC035263.1 | - | 3, 4 | 806 | 4800940 | ||
BC057806.1 | - | 3, 4 | 817 | 30337849 | ||
BT019685.1 | - | 3, 4 | 652 | |||
FJ795026.1 | - | 3, 4 | 651 | |||
M20841.1 | - | 3, 4 | 772 | |||
M31145.1 | - | 3, 4 | 816 | |||
X12385.1 | - | 3, 4 | 802 | |||
X13405.1 | - | 3, 4 | 754 | |||
X15002.1 | - | 2, 3 | 232 | |||
Y00856.1 | - | 3, 4 | 704 |