Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144996.1 | NP_001138468.1 | 23 | 3208 | ||
NM_001144997.1 | NP_001138469.1 | 23 | 2733 | |||
NM_002206.2 | NP_002197.2 | 23 | 3196 | |||
XM_005268839.1 | XP_005268896.1 | 24 | 3461 | |||
XM_005268840.1 | XP_005268897.1 | 24 | 3443 | |||
XM_005268841.1 | XP_005268898.1 | 24 | 3461 | |||
XM_005268842.1 | XP_005268899.1 | 23 | 3311 | |||
XM_005268844.1 | XP_005268901.1 | 25 | 3086 | |||
XM_005268846.1 | XP_005268903.1 | 24 | 2867 | |||
XM_005268848.1 | XP_005268905.1 | 23 | 2885 | |||
XM_005268849.1 | XP_005268906.1 | 23 | 2815 | |||
XM_005268850.1 | XP_005268907.1 | 21 | 2704 | |||
XM_017019265.1 | XP_016874754.1 | 22 | 2789 | |||
GenBank mRNA | AB210036.1 | - | 23 | 3166 | ||
AF032108.1 | - | 23 | 3137 | |||
AF052050.1 | - | 23 | 2976 | |||
AF072132.1 | - | 23 | 2976 | |||
AK022548.1 | - | 21 | 2580 | |||
AK057949.1 | - | 10 | 1041 | |||
AK304864.1 | - | 23 | 2709 | |||
AY358882.1 | - | 23 | 3007 | |||
BC050280.1 | - | 23 | 3192 | 5284443 |