Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105528.1 | NP_001098998.1 | 20 | 2442 | ||
NM_001308126.1 | NP_001295055.1 | 20 | 2442 | |||
NM_198995.2 | NP_945346.2 | 20 | 2397 | |||
XM_011525948.1 | XP_011524250.1 | 21 | 2511 | |||
XM_017025721.1 | XP_016881210.1 | 21 | 2511 | |||
XM_017025722.1 | XP_016881211.1 | 21 | 2532 | |||
XM_017025723.1 | XP_016881212.1 | 21 | 2541 | |||
XM_017025724.1 | XP_016881213.1 | 15 | 2075 | |||
XM_017025725.1 | XP_016881214.1 | 15 | 2692 | |||
GenBank mRNA | BC091517.1 | - | 21 | 2483 | 30389491 | |
BC130535.1 | - | 20 | 2379 | 40146566 | ||
BC144178.1 | - | 20 | 2379 | 9052698 |