Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF568 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204835.1 | NP_001191764.1 | 3 | 538 | ||
NM_001204838.1 | NP_001191767.1 | 3 | 538 | |||
NM_198539.3 | NP_940941.2 | 3 | 538 | |||
XM_017026772.1 | XP_016882261.1 | 3 | 1126 | |||
XM_017026773.1 | XP_016882262.1 | 3 | 529 | |||
XM_017026779.1 | XP_016882268.1 | 3 | 341 | |||
GenBank mRNA | BC016334.1 | - | 3 | 539 | ||
BC031218.1 | - | 3 | 526 | |||
BC108696.1 | - | 3 | 503 | |||
BC111777.1 | - | 3 | 320 | |||
BC131547.1 | - | 3 | 321 | |||
DA010273.1 | - | 3 | 535 |