Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030311.2 | NP_001025482.1 | 13 | 1702 | ||
NM_001030312.2 | NP_001025483.1 | 9 | 1285 | |||
NM_001030313.2 | NP_001025484.1 | 10 | 1417 | |||
NM_001160277.1 | NP_001153749.1 | 12 | 1570 | |||
NM_201548.4 | NP_963842.1 | 12 | 1624 | |||
NR_027689.1 | 10 | 1428 | ||||
NR_027690.1 | 11 | 1560 | ||||
GenBank mRNA | AJ640141.1 | - | 12 | 1541 | ||
AJ697855.1 | - | 10 | 2329 | |||
AJ697856.1 | - | 11 | 1459 | |||
AJ697858.1 | - | 9 | 1317 | |||
AK129976.1 | - | 9 | 867 | |||
AK293844.1 | - | 12 | 1571 | |||
AY357073.1 | - | 12 | 1523 | |||
AY690329.1 | - | 13 | 1601 | |||
AY690330.1 | - | 9 | 1184 | |||
AY690331.1 | - | 10 | 1316 | |||
AY690332.1 | - | 10 | 1327 | |||
AY690333.1 | - | 11 | 1459 | |||
BC137498.1 | - | 12 | 1550 | 9021498 | ||
BC137499.1 | - | 12 | 1550 | 9021499 | ||
HQ426666.1 | - | 12 | 1430 | |||
HQ426667.1 | - | 11 | 1366 | |||
HQ426668.1 | - | 10 | 1234 | |||
HQ426669.1 | - | 10 | 1223 | |||
HQ426670.1 | - | 9 | 1091 | |||
HQ426671.1 | - | 13 | 1430 | |||
HQ426672.1 | - | 12 | 1352 | |||
HQ426673.1 | - | 11 | 1288 | |||
HQ426674.1 | - | 10 | 1156 | |||
HQ426675.1 | - | 10 | 1145 | |||
HQ426676.1 | - | 9 | 1013 | |||
HQ426677.1 | - | 10 | 960 | |||
HQ426678.1 | - | 13 | 1677 | |||
HQ426679.1 | - | 10 | 1982 | |||
JF290420.1 | - | 12 | 1510 |