Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKIDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207371.3 | NP_997254.3 | 4 | 4644 | ||
XM_005252450.3 | XP_005252507.1 | 5 | 3753 | |||
XM_011519480.2 | XP_011517782.1 | 5 | 3750 | |||
XM_017016225.1 | XP_016871714.1 | 5 | 4544 | |||
XM_017016226.1 | XP_016871715.1 | 5 | 4547 | |||
GenBank mRNA | AB234866.1 | - | 2 | 2245 | ||
AK127130.1 | - | 2 | 1679 | |||
AK131456.1 | - | 5 | 4380 | |||
AL834446.1 | - | 1 | 1362 |