Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001669.2 | NP_001001669.2 | 5 | 550 | ||
XM_005268447.4 | XP_005268504.1 | 5 | 521 | |||
XM_005268448.1 | XP_005268505.1 | 4 | 424 | |||
XM_006714784.3 | XP_006714847.1 | 6 | 889 | |||
XM_011537642.2 | XP_011535944.1 | 5 | 572 | |||
XM_017009460.1 | XP_016864949.1 | 5 | 774 | |||
XM_017009461.1 | XP_016864950.1 | 4 | 424 | |||
GenBank mRNA | AB199796.1 | - | 4 | 497 | ||
AK300355.1 | - | 4 | 411 | |||
AK300559.1 | - | 5 | 649 | |||
AK316002.1 | - | 5 | 620 | |||
BC041325.1 | - | 5 | 550 |