Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AFF3 SIRNA Assays ›
Gene Symbol:
AFF3
Gene Name:
AF4/FMR2 family member 3
Gene Aliases:
LAF4, MLLT2-like
Chromosome Location:
Chr.2: 99545861 - 100142583 on Build GRCh38
UniGene ID:
Hs.444414
Species:
Human
siRNA ID
HSS180451
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AFF3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025108.1
NP_001020279.1
21, 22
3502
NM_002285.2
NP_002276.2
21, 22
3519
XM_005263943.3
XP_005264000.2
20, 21
3396
XM_005263945.3
XP_005264002.1
12, 13
2753
XM_011511169.2
XP_011509471.1
21, 22
3499
XM_011511170.2
XP_011509472.1
22, 23
3638
XM_011511171.2
XP_011509473.1
21, 22
3502
XM_011511172.2
XP_011509474.1
20, 21
3454
XM_011511173.2
XP_011509475.1
21, 22
3515
XM_011511174.2
XP_011509476.1
21, 22
3459
XM_011511175.2
XP_011509477.1
23, 24
3749
XM_011511176.2
XP_011509478.1
20, 21
3440
XM_011511177.2
XP_011509479.1
21, 22
3530
XM_011511178.2
XP_011509480.1
11, 12
2261
XM_011511179.2
XP_011509481.1
11, 12
2258
XM_017004085.1
XP_016859574.1
12, 13
2750
XM_017004086.1
XP_016859575.1
12, 13
3231
XM_017004087.1
XP_016859576.1
12, 13
3228
GenBank mRNA
AB209634.1
-
8, 9
1221
BC136579.1
-
19, 20
3302
9020569
BC144266.1
-
20, 21
3377
9052786
CN409428.1
-
3, 4
295
U34360.1
-
19, 20
3340
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
response to tumor necrosis factor
embryonic hindlimb morphogenesis
double-stranded DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0