Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291516.1 | NP_001278445.1 | 3 | 601 | ||
NR_111982.1 | 3 | 918 | ||||
XM_011518679.2 | XP_011516981.1 | 3 | 924 | |||
XM_011518681.2 | XP_011516983.1 | 3 | 923 | |||
XM_011518683.2 | XP_011516985.1 | 3 | 923 | |||
XM_011518684.2 | XP_011516986.1 | 3 | 923 | |||
GenBank mRNA | AK125773.1 | - | 3 | 917 | ||
BC031926.1 | - | 3 | 601 | |||
BC042107.1 | - | 3 | 894 | 5170879 |