Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164478.1 | NP_001157950.1 | 3 | 195 | ||
NM_001164479.1 | NP_001157951.1 | 3 | 195 | |||
GenBank mRNA | AK126569.1 | - | 3 | 183 | ||
BQ269742.1 | - | 3 | 514 | |||
DA507232.1 | - | 3 | 195 |