Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002318.2 | NP_002309.1 | 7 | 1534 | ||
GenBank mRNA | AF117949.1 | - | 4 | 664 | ||
AK222477.1 | - | 7 | 1445 | |||
AK293292.1 | - | 4 | 360 | |||
AK312266.1 | - | 7 | 1446 | |||
BC000594.2 | - | 7 | 1404 | 3347512 | ||
BC020481.1 | - | 7 | 1454 | |||
KF928961.1 | - | 6 | 1195 | |||
KF975332.1 | - | 6 | 1135 | |||
U89942.1 | - | 7 | 1442 |