Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGC57346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026680.3 | 4 | 489 | |||
NR_027295.2 | 4 | 983 | ||||
GenBank mRNA | AK311451.1 | - | 4 | 472 | ||
BC045575.1 | - | 4 | 988 |