Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC403323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122077.1 | 2, 3 | 787 | |||
GenBank mRNA | AK000451.1 | - | 2, 3 | 787 | ||
DB521113.1 | - | 2, 3 | 328 |
Gene Symbol: XLOC_007697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131213.1 | 1, 2 | 31 |