Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002349.3 | NP_002340.2 | 18 | 2610 | ||
GenBank mRNA | AB208915.1 | - | 12 | 1390 | ||
AF064827.1 | - | 18 | 2536 | |||
DB146492.1 | - | 3 | 213 |
Gene Symbol: LY75-CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198759.1 | NP_001185688.1 | 18 | 2610 | ||
NM_001198760.1 | NP_001185689.1 | 18 | 2610 |