Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP4 SIRNA Assays ›
Gene Symbol:
MAP4
Gene Name:
microtubule associated protein 4
Gene Aliases:
-
Chromosome Location:
Chr.3: 47850690 - 48089279 on Build GRCh38
UniGene ID:
Hs.517949
Species:
Human
siRNA ID
HSS181002
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134364.1
NP_001127836.1
7
2084
NM_002375.4
NP_002366.2
7
2084
XM_005265133.4
XP_005265190.1
8
1715
XM_005265134.4
XP_005265191.1
8
2128
XM_005265155.4
XP_005265212.1
6
2008
XM_006713146.3
XP_006713209.1
9
1747
XM_006713147.3
XP_006713210.1
9
2164
XM_006713152.3
XP_006713215.1
8
2113
XM_011533703.2
XP_011532005.1
8
1664
XM_011533704.2
XP_011532006.1
8
1664
XM_011533705.1
XP_011532007.1
8
1664
XM_011533708.2
XP_011532010.1
8
1664
XM_011533709.2
XP_011532011.1
8
1664
XM_011533710.2
XP_011532012.1
7
1541
XM_011533718.1
XP_011532020.1
8
1664
XM_011533719.2
XP_011532021.1
8
1664
XM_017006391.1
XP_016861880.1
8
1664
XM_017006392.1
XP_016861881.1
8
1664
XM_017006393.1
XP_016861882.1
8
1696
XM_017006394.1
XP_016861883.1
7
1990
XM_017006395.1
XP_016861884.1
8
1664
XM_017006396.1
XP_016861885.1
8
1664
XM_017006397.1
XP_016861886.1
8
1664
XM_017006398.1
XP_016861887.1
7
1613
XM_017006399.1
XP_016861888.1
8
2128
XM_017006400.1
XP_016861889.1
8
2128
XM_017006401.1
XP_016861890.1
6
1505
XM_017006402.1
XP_016861891.1
7
2077
XM_017006403.1
XP_016861892.1
8
1664
XM_017006404.1
XP_016861893.1
8
1664
XM_017006405.1
XP_016861894.1
7
2077
XM_017006406.1
XP_016861895.1
8
2128
XM_017006407.1
XP_016861896.1
8
1664
XM_017006408.1
XP_016861897.1
8
1664
XM_017006409.1
XP_016861898.1
7
2077
XM_017006410.1
XP_016861899.1
7
2005
XM_017006411.1
XP_016861900.1
6
1954
XM_017006412.1
XP_016861901.1
6
1954
XM_017006413.1
XP_016861902.1
7
2077
XM_017006414.1
XP_016861903.1
7
2077
XM_017006415.1
XP_016861904.1
7
2077
XM_017006416.1
XP_016861905.1
5
1454
GenBank mRNA
AB209377.1
-
7
1644
AK297242.1
-
6
1583
AK307606.1
-
8
1697
BC008715.2
-
7
1648
2961360
BC012794.2
-
7
1648
4098499
BC015149.2
-
7
1648
2961360
M64571.1
-
7
1648
U19727.1
-
7
1633
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mitotic spindle organization
neuron projection development
microtubule sliding
establishment of spindle orientation
cell division
negative regulation of nonmotile primary cilium assembly
structural molecule activity
protein binding
microtubule binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0