Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP4 SIRNA Assays ›
Gene Symbol:
MAP4
Gene Name:
microtubule associated protein 4
Gene Aliases:
-
Chromosome Location:
Chr.3: 47850690 - 48089279 on Build GRCh38
UniGene ID:
Hs.517949
Species:
Human
siRNA ID
HSS181003
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134364.1
NP_001127836.1
16
3612
NM_002375.4
NP_002366.2
16
3612
XM_005265133.4
XP_005265190.1
19
6678
XM_005265134.4
XP_005265191.1
19
7091
XM_005265155.4
XP_005265212.1
15
3536
XM_005265157.4
XP_005265214.1
9
2905
XM_005265158.2
XP_005265215.1
7
2698
XM_006713146.3
XP_006713209.1
20
6710
XM_006713147.3
XP_006713210.1
20
7127
XM_006713152.3
XP_006713215.1
19
7076
XM_011533703.2
XP_011532005.1
19
6627
XM_011533704.2
XP_011532006.1
19
6582
XM_011533705.1
XP_011532007.1
18
6576
XM_011533708.2
XP_011532010.1
18
6531
XM_011533709.2
XP_011532011.1
18
6513
XM_011533710.2
XP_011532012.1
18
6504
XM_011533718.1
XP_011532020.1
16
6369
XM_011533719.2
XP_011532021.1
17
3192
XM_017006391.1
XP_016861880.1
19
6582
XM_017006392.1
XP_016861881.1
18
6513
XM_017006393.1
XP_016861882.1
19
6659
XM_017006394.1
XP_016861883.1
18
6953
XM_017006395.1
XP_016861884.1
17
3192
XM_017006396.1
XP_016861885.1
17
3147
XM_017006397.1
XP_016861886.1
17
3147
XM_017006398.1
XP_016861887.1
16
3141
XM_017006399.1
XP_016861888.1
17
3656
XM_017006400.1
XP_016861889.1
17
3656
XM_017006401.1
XP_016861890.1
15
3033
XM_017006402.1
XP_016861891.1
16
3605
XM_017006403.1
XP_016861892.1
15
2985
XM_017006404.1
XP_016861893.1
15
2985
XM_017006405.1
XP_016861894.1
16
3560
XM_017006406.1
XP_016861895.1
16
3542
XM_017006407.1
XP_016861896.1
15
2940
XM_017006408.1
XP_016861897.1
15
2940
XM_017006409.1
XP_016861898.1
15
3491
XM_017006410.1
XP_016861899.1
16
3488
XM_017006411.1
XP_016861900.1
15
3482
XM_017006412.1
XP_016861901.1
15
3482
XM_017006413.1
XP_016861902.1
14
3398
XM_017006414.1
XP_016861903.1
14
3398
XM_017006415.1
XP_016861904.1
14
3353
XM_017006416.1
XP_016861905.1
12
2775
XM_017006417.1
XP_016861906.1
9
2905
XM_017006418.1
XP_016861907.1
9
2860
XM_017006419.1
XP_016861908.1
8
2791
XM_017006420.1
XP_016861909.1
8
2791
XM_017006421.1
XP_016861910.1
7
2698
XM_017006422.1
XP_016861911.1
7
2653
GenBank mRNA
AB209377.1
-
16
3172
AK299854.1
-
7
1039
AL831878.1
-
3
191
BC008715.2
-
14
2657
2961360
BC012794.2
-
14
2657
4098499
BC015149.2
-
14
2657
2961360
BC048200.1
-
9
2203
CR749544.1
-
7
1110
M64571.1
-
16
3176
U19727.1
-
16
3161
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mitotic spindle organization
neuron projection development
microtubule sliding
establishment of spindle orientation
cell division
negative regulation of nonmotile primary cilium assembly
structural molecule activity
protein binding
microtubule binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0