Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197320.1 | NP_001184249.1 | 4 | 700 | ||
NM_021960.4 | NP_068779.1 | 3 | 1159 | |||
NM_182763.2 | NP_877495.1 | 2 | 911 | |||
GenBank mRNA | AF118124.1 | - | 3 | 1000 | ||
AF203373.1 | - | 2 | 703 | |||
AK294462.1 | - | 4 | 731 | |||
AK297217.1 | - | 4 | 695 | |||
AK300499.1 | - | 4 | 805 | |||
AK304775.1 | - | 4 | 841 | |||
AK312508.1 | - | 3 | 1031 | |||
AK316267.1 | - | 4 | 987 | |||
BC017197.2 | - | 3 | 1005 | 3138465 | ||
BC071897.1 | - | 3 | 1007 | 6586159 | ||
BC107735.1 | - | 3 | 1022 | 4770487 | ||
BT006640.1 | - | 3 | 951 | |||
L08246.1 | - | 3 | 1011 |