Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CD46 SIRNA Assays ›
Gene Symbol:
CD46
Gene Name:
CD46 molecule
Gene Aliases:
AHUS2, MCP, MIC10, TLX, TRA2.10
Chromosome Location:
Chr.1: 207752038 - 207795516 on Build GRCh38
UniGene ID:
Hs.510402
Species:
Human
siRNA ID
HSS181050
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CD46
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002389.4
NP_002380.3
3
523
NM_153826.3
NP_722548.1
3
523
NM_172350.2
NP_758860.1
3
523
NM_172351.2
NP_758861.1
3
523
NM_172352.2
NP_758862.1
3
523
NM_172353.2
NP_758863.1
3
523
NM_172359.2
NP_758869.1
3
523
NM_172361.2
NP_758871.1
3
523
XM_011509563.1
XP_011507865.1
3
523
XM_011509564.1
XP_011507866.1
3
523
XM_017001308.1
XP_016856797.1
3
523
XM_017001309.1
XP_016856798.1
3
523
XM_017001310.1
XP_016856799.1
3
523
GenBank mRNA
A18585.1
-
3
381
AF209712.1
-
3
348
AF209713.1
-
3
348
AF209714.1
-
3
348
AK222822.1
-
3
449
AK291227.1
-
3
477
AK296819.1
-
3
475
AU131238.2
-
3
504
BC030594.2
-
3
463
4823040
BX537451.1
-
3
494
BX640613.1
-
3
505
BX649050.1
-
3
494
D84105.1
-
3
369
DB078198.1
-
3
523
EF076055.1
-
3
381
EF076056.1
-
3
381
EF076057.1
-
3
381
EF076058.1
-
3
381
M58050.1
-
3
382
S51940.1
-
3
435
X59405.1
-
3
493
X59406.1
-
3
452
X59407.1
-
3
452
X59408.1
-
3
493
X59409.1
-
3
452
X59410.1
-
3
452
Y00651.1
-
3
391
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
T cell mediated immunity
proteolysis
complement activation, classical pathway
single fertilization
regulation of Notch signaling pathway
positive regulation of gene expression
negative regulation of gene expression
regulation of complement activation
interleukin-10 production
positive regulation of interleukin-10 production
sequestering of extracellular ligand from receptor
positive regulation of T cell proliferation
negative regulation of catalytic activity
positive regulation of memory T cell differentiation
innate immune response
positive regulation of regulatory T cell differentiation
negative regulation of complement activation
viral entry into host cell
positive regulation of transforming growth factor beta production
virus receptor activity
complement binding
endopeptidase activity
enzyme inhibitor activity
receptor activity
protein binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0