Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300804.1 | NP_001287733.1 | 6 | 1061 | ||
NM_001300805.1 | NP_001287734.1 | 4 | 741 | |||
NM_001300806.1 | NP_001287735.1 | 4 | 1010 | |||
NM_005586.3 | NP_005577.1 | 5 | 924 | |||
XM_005249117.3 | XP_005249174.1 | 5 | 927 | |||
XM_011514625.2 | XP_011512927.1 | 6 | 1212 | |||
XM_011514626.2 | XP_011512928.1 | 5 | 1035 | |||
XM_017010867.1 | XP_016866356.1 | 4 | 1583 | |||
GenBank mRNA | AK290735.1 | - | 4 | 1008 | ||
BC007836.2 | - | 5 | 873 | 4110841 | ||
BI824391.1 | - | 4 | 721 | |||
U78313.1 | - | 5 | 866 |