Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318942.1 | NP_001305871.1 | 3 | 436 | ||
NM_001318943.1 | NP_001305872.1 | 3 | 817 | |||
NM_014623.3 | NP_055438.1 | 3 | 479 | |||
XM_005249121.3 | XP_005249178.1 | 3 | 544 | |||
XM_017010868.1 | XP_016866357.1 | 3 | 375 | |||
GenBank mRNA | AA448322.1 | - | 2 | 427 | ||
AK309759.1 | - | 3 | 377 | |||
BC001754.1 | - | 3 | 437 | 3355279 | ||
BM450289.1 | - | 380 | ||||
BM552852.1 | - | 3 | 795 | |||
BM969076.1 | - | 2 | 570 | |||
BQ232807.1 | - | 3 | 479 | |||
BT009831.1 | - | 3 | 378 | |||
L10400.1 | - | 3 | 412 | |||
M27937.1 | - | 3 | 412 |