Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATXN3 SIRNA Assays ›
Gene Symbol:
ATXN3
ATXN8
Gene Name:
ataxin 3
ataxin 8
Gene Aliases:
AT3, ATX3, JOS, MJD, MJD1, SCA3
ATXN3
Chromosome Location:
Chr.14: 92058552 - 92106621 on Build GRCh38
Chromosome Location:
-
UniGene ID:
Hs.532632
Hs.532632
Species:
Human
siRNA ID
HSS181112
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127696.1
NP_001121168.1
8
839
NM_001127697.2
NP_001121169.2
7
731
NM_001164779.1
NP_001158251.1
5
521
NM_001164780.1
NP_001158252.1
6
588
NM_001164781.1
NP_001158253.1
7
674
NM_004993.5
NP_004984.2
9
884
NM_030660.4
NP_109376.1
8
719
NR_028453.1
8
798
NR_028454.1
7
633
NR_028455.1
8
798
NR_028456.1
7
633
NR_028457.1
9
889
NR_028458.1
8
775
NR_028459.1
9
884
NR_028461.1
8
796
NR_028462.1
8
775
NR_028463.1
4
433
NR_028464.1
7
731
NR_028465.1
7
753
NR_028466.1
4
433
NR_028467.1
8
799
NR_028468.1
7
631
NR_028469.1
7
633
NR_031765.1
3
300
GenBank mRNA
AB050194.1
-
8
708
AB209309.1
-
2
160
AK225884.1
-
9
823
AK307720.1
-
6
549
AK314919.1
-
9
836
BC022245.1
-
8
767
BC033711.1
-
9
827
4393766
BC095402.1
-
9
844
30347215
BU190081.1
-
8
818
GQ176433.1
-
9
837
GQ176434.1
-
9
836
GQ176435.1
-
9
837
GQ176436.1
-
7
684
GQ176437.1
-
7
684
GQ176438.1
-
8
751
GQ176440.1
-
8
672
GQ176441.1
-
8
751
GQ176443.1
-
8
751
GQ176445.1
-
7
586
GQ176446.1
-
7
583
GQ176448.1
-
9
837
GQ176451.1
-
7
586
GQ176452.1
-
8
837
GQ176453.1
-
8
727
GQ176454.1
-
9
837
GQ176455.1
-
8
672
GQ176456.1
-
7
586
GQ176457.1
-
7
684
GQ176458.1
-
8
751
GQ176459.1
-
5
474
GQ176462.1
-
9
837
GQ176463.1
-
8
751
GQ176464.1
-
8
751
GQ176465.1
-
4
386
GQ176466.1
-
7
586
GQ176469.1
-
7
684
GQ176470.1
-
7
706
GQ176474.1
-
7
684
GQ176475.1
-
7
586
GQ176476.1
-
9
837
GQ176477.1
-
8
751
GQ176480.1
-
7
584
GQ176481.1
-
8
751
GQ176483.1
-
8
751
GQ176484.1
-
8
751
GQ176485.1
-
7
683
GQ176486.1
-
8
792
GQ176487.1
-
7
684
GQ176488.1
-
9
837
GQ176489.1
-
8
792
GQ176490.1
-
7
718
GQ176492.1
-
8
751
GQ176493.1
-
7
683
GQ176495.1
-
7
684
GQ176496.1
-
7
586
GQ176497.1
-
8
751
GQ176498.1
-
8
672
GQ176499.1
-
9
837
GQ176500.1
-
9
838
GQ176501.1
-
8
728
GQ176503.1
-
7
585
GQ176504.1
-
9
837
GQ176505.1
-
7
586
GQ176506.1
-
7
586
GQ176507.1
-
8
750
GQ176508.1
-
9
837
GQ176510.1
-
9
837
GQ176511.1
-
7
684
GQ176513.1
-
8
751
GQ176514.1
-
6
541
GQ176515.1
-
7
584
GQ176516.1
-
9
837
GQ176517.1
-
4
386
GQ176518.1
-
4
386
GQ176519.1
-
8
750
GQ176520.1
-
6
541
GQ176521.1
-
7
586
GQ176522.1
-
7
586
GQ176524.1
-
8
751
GQ176525.1
-
7
586
GQ176527.1
-
8
751
GQ176528.1
-
8
751
GQ176529.1
-
8
748
GQ176530.1
-
8
751
GQ176531.1
-
9
837
GQ176532.1
-
8
837
GQ176533.1
-
9
837
GQ176534.1
-
8
748
GQ176535.1
-
586
GQ176536.1
-
8
672
GQ176537.1
-
8
751
GQ176538.1
-
9
837
GQ176539.1
-
8
672
GQ176540.1
-
8
725
GQ176542.1
-
5
473
GQ176543.1
-
7
586
GQ176544.1
-
7
586
GQ176545.1
-
5
474
GQ176546.1
-
8
751
GQ176547.1
-
7
586
GQ176548.1
-
9
837
GQ176549.1
-
7
586
GQ176550.1
-
9
837
GQ176551.1
-
9
835
GQ176552.1
-
9
836
GQ176553.1
-
7
584
GQ176554.1
-
7
586
GQ176556.1
-
7
586
GQ176558.1
-
7
584
GQ176559.1
-
7
586
GQ176560.1
-
9
837
GQ176563.1
-
9
837
GQ176565.1
-
9
837
GQ176566.1
-
8
672
GQ176567.1
-
9
837
GQ176568.1
-
9
837
GQ176569.1
-
8
748
GQ176570.1
-
9
837
GQ176571.1
-
7
586
GQ176572.1
-
8
751
GQ176573.1
-
8
672
GQ176577.1
-
9
837
GQ176578.1
-
9
837
GQ176579.1
-
8
728
GQ176580.1
-
9
837
GQ176583.1
-
3
253
GQ176585.1
-
7
585
GQ176586.1
-
8
751
GQ176587.1
-
7
586
GQ176588.1
-
8
751
GQ176589.1
-
9
837
GQ176590.1
-
3
253
GQ176591.1
-
7
586
GQ176592.1
-
8
671
GQ176593.1
-
10
923
GQ176594.1
-
9
837
GQ176595.1
-
7
584
GQ176596.1
-
8
751
GQ176597.1
-
9
837
GQ176598.1
-
7
586
GQ176599.1
-
9
837
GQ176600.1
-
8
751
GQ176601.1
-
7
586
GQ176602.1
-
9
855
GQ176603.1
-
7
586
GQ176604.1
-
7
586
GQ176605.1
-
8
751
GQ176607.1
-
8
751
GQ176608.1
-
7
684
GQ176610.1
-
8
751
GQ176611.1
-
8
751
GQ176612.1
-
9
837
GQ176614.1
-
6
541
GQ176617.1
-
8
751
GQ176619.1
-
9
837
GQ176620.1
-
8
751
GQ176621.1
-
9
837
GQ176622.1
-
9
837
GQ176623.1
-
7
586
GQ176624.1
-
8
770
GQ176625.1
-
474
GQ176626.1
-
7
586
GQ176627.1
-
5
474
GQ176628.1
-
837
GQ176630.1
-
8
751
GQ176631.1
-
7
684
GQ176632.1
-
9
837
GQ176633.1
-
5
474
GQ176635.1
-
586
GQ176636.1
-
6
658
GQ176637.1
-
7
586
GQ176639.1
-
7
683
GQ176640.1
-
9
837
GQ176641.1
-
557
GQ176642.1
-
9
837
GQ176643.1
-
586
GQ176644.1
-
9
832
GQ176645.1
-
9
837
GQ176646.1
-
9
837
GQ176647.1
-
586
GQ176648.1
-
8
765
GQ176649.1
-
8
751
GQ176650.1
-
4
386
GQ176653.1
-
6
541
GQ176654.1
-
8
751
GQ176655.1
-
7
681
GQ176657.1
-
7
586
GQ176658.1
-
9
837
GQ176659.1
-
751
GQ176660.1
-
8
751
GQ176661.1
-
6
541
GQ176662.1
-
8
751
GQ176663.1
-
7
684
GQ176664.1
-
8
751
GQ176665.1
-
8
672
GQ176666.1
-
6
541
GQ176668.1
-
9
834
GQ176669.1
-
751
GQ176670.1
-
6
639
GQ176671.1
-
837
GQ176673.1
-
9
837
GQ176674.1
-
9
837
GQ176675.1
-
8
672
GQ176676.1
-
7
586
GQ176677.1
-
9
837
GQ176678.1
-
8
751
GQ176679.1
-
8
672
GQ176680.1
-
9
837
GQ176681.1
-
6
596
GQ176682.1
-
8
751
GQ176685.1
-
9
836
GQ176687.1
-
9
837
GQ176688.1
-
7
683
GQ176689.1
-
6
541
GQ176690.1
-
8
751
GQ176691.1
-
7
586
GQ176692.1
-
9
837
GQ176693.1
-
837
GQ176694.1
-
7
505
GQ176695.1
-
7
684
GQ176696.1
-
8
751
GQ176697.1
-
9
837
GQ176699.1
-
837
GQ176700.1
-
8
672
GQ176702.1
-
6
541
GQ176703.1
-
9
837
GQ176705.1
-
5
474
GQ176707.1
-
583
GQ176708.1
-
10
923
GQ176709.1
-
748
GQ176711.1
-
6
541
GQ176712.1
-
8
792
GQ176713.1
-
586
GQ176714.1
-
9
837
GQ176715.1
-
9
759
GQ176716.1
-
7
586
GQ176717.1
-
751
GQ176719.1
-
837
GQ176720.1
-
7
586
GQ176721.1
-
9
837
GQ176722.1
-
8
751
GQ176723.1
-
6
541
GQ176724.1
-
7
684
GQ176725.1
-
9
837
GQ176726.1
-
7
586
GQ176727.1
-
5
474
GQ176728.1
-
9
837
GQ176729.1
-
7
582
GQ176731.1
-
7
586
GQ176732.1
-
586
GQ176733.1
-
8
751
GQ176734.1
-
684
GQ176735.1
-
9
837
GQ176736.1
-
586
GQ176737.1
-
9
837
GQ176738.1
-
8
751
GQ176740.1
-
7
586
GQ176741.1
-
837
GQ176742.1
-
519
GQ176743.1
-
7
586
GQ176744.1
-
8
792
GQ176745.1
-
837
GQ176747.1
-
837
GQ176748.1
-
7
586
GQ176749.1
-
9
837
GQ176750.1
-
8
672
GQ176751.1
-
586
GQ176753.1
-
6
541
GQ176754.1
-
8
672
GQ176755.1
-
8
751
GQ176756.1
-
9
837
GQ176757.1
-
7
684
GQ176758.1
-
7
586
GQ176759.1
-
9
837
GQ176760.1
-
8
751
GQ176761.1
-
6
519
GQ176762.1
-
837
GQ176763.1
-
9
837
GQ176764.1
-
586
GQ176765.1
-
7
586
GQ176766.1
-
9
837
GQ176767.1
-
8
751
GQ176769.1
-
6
639
GQ176770.1
-
6
419
GQ176771.1
-
9
837
GQ176772.1
-
7
684
GQ176774.1
-
7
586
GQ176778.1
-
6
541
GQ176779.1
-
7
684
GQ176780.1
-
7
586
GQ176781.1
-
7
684
GQ176782.1
-
7
684
GQ176783.1
-
586
GQ176784.1
-
7
684
GQ176785.1
-
6
519
GQ176787.1
-
9
837
GQ176788.1
-
7
586
GQ176789.1
-
7
683
GQ176791.1
-
9
837
GQ176792.1
-
7
681
GQ176793.1
-
7
684
GQ176794.1
-
541
GQ176795.1
-
7
586
GQ176796.1
-
751
GQ176797.1
-
8
751
GQ176798.1
-
751
GQ176799.1
-
519
GQ176801.1
-
750
GQ176802.1
-
684
GQ176803.1
-
751
GQ176805.1
-
9
813
GQ176806.1
-
9
836
GQ176807.1
-
8
751
GQ176808.1
-
837
GQ176809.1
-
6
635
GQ176810.1
-
8
749
GQ176812.1
-
7
684
GQ176813.1
-
8
672
GQ176814.1
-
7
583
GQ176815.1
-
7
685
GQ176817.1
-
5
473
GQ176818.1
-
8
792
GQ176819.1
-
8
751
GQ176820.1
-
9
837
GQ176821.1
-
7
585
GQ176822.1
-
8
672
GQ176823.1
-
8
728
GQ176824.1
-
9
837
GQ176827.1
-
9
837
GQ176829.1
-
9
837
GQ176831.1
-
837
GQ176832.1
-
7
586
GQ176833.1
-
8
747
GQ176834.1
-
750
GQ176835.1
-
8
671
GQ176836.1
-
8
671
GQ176837.1
-
9
837
GQ176838.1
-
9
837
GQ176839.1
-
582
GQ176840.1
-
7
586
GQ176841.1
-
9
837
GQ176842.1
-
751
GQ176843.1
-
7
585
GQ176844.1
-
7
599
GQ176845.1
-
8
750
GQ176846.1
-
9
837
GQ176847.1
-
7
583
S75313.1
-
9
850
U64820.1
-
9
872
U64821.1
-
9
835
U64822.1
-
8
812
Gene Symbol: ATXN8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
GQ176777.1
-
519
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
nucleotide-excision repair
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
misfolded or incompletely synthesized protein catabolic process
chemical synaptic transmission
nervous system development
regulation of cell-substrate adhesion
protein deubiquitination
actin cytoskeleton organization
cellular response to heat
monoubiquitinated protein deubiquitination
exploration behavior
proteasome-mediated ubiquitin-dependent protein catabolic process
intermediate filament cytoskeleton organization
protein K63-linked deubiquitination
histone H3 deacetylation
protein K48-linked deubiquitination
cellular response to misfolded protein
positive regulation of ERAD pathway
protein localization to cytosolic proteasome complex involved in ERAD pathway
RNA polymerase II regulatory region DNA binding
histone deacetylase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
omega peptidase activity
ubiquitin protein ligase binding
identical protein binding
ATPase binding
Lys63-specific deubiquitinase activity
Lys48-specific deubiquitinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0