Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272101.1 | NP_001259030.1 | 1, 2 | 243 | ||
NM_002429.5 | NP_002420.1 | 1, 2 | 243 | |||
NR_073606.1 | 1, 2 | 243 | ||||
GenBank mRNA | AK225939.1 | - | 1, 2 | 233 | ||
AK297999.1 | - | 1, 2 | 200 | |||
AK303202.1 | - | 1, 2 | 196 | |||
AL545199.3 | - | 1, 2 | 243 | |||
BC050368.1 | - | 1, 2 | 182 | 5761074 | ||
BI761698.1 | - | 1, 2 | 181 | |||
U37791.1 | - | 1, 2 | 171 | |||
U38321.1 | - | 1, 2 | 166 | |||
U38431.1 | - | 1, 2 | 164 | |||
X92521.1 | - | 1, 2 | 191 | |||
Y08622.1 | - | 1, 2 | 190 |