Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
siRNA ID
HSS181370
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
7
702
NM_001018017.2
NP_001018017.1
7
675
NM_001044390.2
NP_001037855.1
6
519
NM_001044391.2
NP_001037856.1
4
360
NM_001044392.2
NP_001037857.1
4
387
NM_001044393.2
NP_001037858.1
6
487
NM_001204285.1
NP_001191214.1
7
1335
NM_001204286.1
NP_001191215.1
7
1362
NM_001204287.1
NP_001191216.1
7
756
NM_001204288.1
NP_001191217.1
7
670
NM_001204289.1
NP_001191218.1
6
624
NM_001204290.1
NP_001191219.1
6
561
NM_001204291.1
NP_001191220.1
6
633
NM_001204292.1
NP_001191221.1
6
627
NM_001204293.1
NP_001191222.1
6
607
NM_001204294.1
NP_001191223.1
6
600
NM_001204295.1
NP_001191224.1
5
477
NM_001204296.1
NP_001191225.1
6
546
NM_001204297.1
NP_001191226.1
4
441
NM_002456.5
NP_002447.4
7
729
GenBank mRNA
AF125525.1
-
7
594
AF348143.1
-
4
366
AK301575.1
-
8
1587
AY327582.1
-
6
471
AY327583.1
-
4
312
AY327584.1
-
4
339
AY327585.1
-
4
333
AY327586.1
-
4
360
AY327587.1
-
7
680
AY327588.1
-
6
575
AY327589.1
-
4
365
AY327590.1
-
5
415
AY327591.1
-
6
470
AY327592.1
-
6
490
AY327595.1
-
5
402
AY327596.1
-
8
810
AY327597.1
-
5
349
AY327598.1
-
6
439
AY327599.1
-
5
442
AY327600.1
-
6
458
BC120974.2
-
7
652
40115473
BC120975.2
-
7
679
40115475
BM969296.1
-
2
489
BM971650.1
-
2
489
EF583642.1
-
7
1005
EF583643.1
-
7
833
EF583644.1
-
7
819
EF583645.1
-
7
779
EF583646.1
-
7
747
EF583647.1
-
7
734
EF583648.1
-
7
661
EF583649.1
-
7
648
EF583650.1
-
7
617
EF583651.1
-
7
612
EF583652.1
-
6
602
EF583653.1
-
6
560
EF583654.1
-
6
539
EF583655.1
-
5
487
EF583656.1
-
4
348
EF670706.1
-
7
1029
EF670707.1
-
7
698
EF670708.1
-
6
611
EF670709.1
-
6
605
EF670710.1
-
6
597
EF670711.1
-
6
585
EF670712.1
-
6
578
EF670713.1
-
6
543
EF670714.1
-
6
533
EF670715.1
-
4
527
EF670716.1
-
5
455
EF670717.1
-
4
365
EF670718.1
-
4
419
EF670719.1
-
4
389
EF670720.1
-
3
349
EF670721.1
-
4
346
EF670722.1
-
4
323
FJ226028.1
-
6
1136
FJ226029.1
-
6
1294
FJ226030.1
-
7
1264
FJ226031.1
-
6
1222
FJ226032.1
-
6
1155
FJ226033.1
-
6
1136
FJ226034.1
-
7
1055
FJ226035.1
-
6
543
FJ226036.1
-
5
1018
FJ226037.1
-
7
833
FJ226038.1
-
4
961
FJ226039.1
-
7
760
FJ226040.1
-
7
680
FJ226041.1
-
8
837
FJ226042.1
-
7
734
FJ226043.1
-
7
720
FJ226044.1
-
6
704
FJ226045.1
-
6
674
FJ226046.1
-
6
669
FJ226047.1
-
7
634
FJ226049.1
-
6
597
FJ226050.1
-
6
570
FJ226051.1
-
6
543
FJ226052.1
-
6
531
FJ226053.1
-
6
524
FJ226054.1
-
5
520
J05581.1
-
7
1335
J05582.1
-
3676
M34088.1
-
5
822
M34089.1
-
5
822
U60259.1
-
6
553
U60260.1
-
7
628
U60261.1
-
7
682
X52228.1
-
4
1913
X52229.1
-
8
1440
X80761.1
-
7
603
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline