Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080527.1 | NP_001073996.1 | 29 | 3959 | ||
XM_006712539.3 | XP_006712602.1 | 30 | 4150 | |||
XM_011511218.2 | XP_011509520.1 | 30 | 4159 | |||
XM_017004169.1 | XP_016859658.1 | 30 | 4159 | |||
GenBank mRNA | AB290182.1 | - | 5 | 813 | ||
AK074183.1 | - | 4 | 891 | |||
CR627350.1 | - | 759 |