Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322019.1 | NP_001308948.1 | 9 | 1006 | ||
NM_004544.3 | NP_004535.1 | 9 | 1006 | |||
NR_136155.1 | 8 | 4048 | ||||
NR_136157.1 | 7 | 3879 | ||||
XM_011511228.2 | XP_011509530.1 | 9 | 993 | |||
GenBank mRNA | AB209440.1 | - | 8 | 4096 | ||
AF087661.1 | - | 9 | 926 | |||
AF087854.1 | - | 9 | 945 | |||
AF453834.1 | - | 10 | 995 | |||
AK091719.1 | - | 4 | 2007 | |||
AK290778.1 | - | 9 | 928 | |||
BC003417.1 | - | 9 | 940 | 3451514 |