Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002497.3 | NP_002488.1 | 8 | 1368 | ||
XM_005273147.1 | XP_005273204.1 | 8 | 1341 | |||
GenBank mRNA | AK223353.1 | - | 8 | 1329 | ||
AK303092.1 | - | 9 | 1231 | |||
AY863109.1 | - | 4 | 548 | |||
BC043502.1 | - | 8 | 1356 | 5541026 | ||
BT019729.1 | - | 8 | 1217 | |||
DQ786247.1 | - | 2 | 249 | |||
DQ786281.1 | - | 2 | 205 | |||
U11050.1 | - | 8 | 1351 | |||
Z29066.1 | - | 8 | 1299 |