Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286075.1 | NP_001273004.1 | 19 | 2739 | ||
NM_004320.4 | NP_004311.1 | 21 | 3090 | |||
NM_173201.3 | NP_775293.1 | 21 | 3090 | |||
GenBank mRNA | AK092691.1 | - | 7 | 887 | ||
AK128456.1 | - | 19 | 2729 | |||
AK291314.1 | - | 21 | 3019 | |||
BC104452.2 | - | 3 | 127 | |||
BC104453.2 | - | 3 | 127 | |||
BU737186.1 | - | 2 | 376 | |||
BX537389.1 | - | 21 | 3086 | |||
BX537784.1 | - | 21 | 3088 | |||
BX647365.1 | - | 21 | 3086 | |||
BX647366.1 | - | 21 | 3082 | |||
BX647367.1 | - | 21 | 3090 | |||
BX647368.1 | - | 14 | 2126 | |||
BX649072.1 | - | 21 | 3078 |